Tisk…

Ethyl 3,6-dichloroimidazo[1,2-a]pyridine-2-carboxylate

Dodavatel: Apollo Scientific
Varování

OR110199-1G OR110199-5G
APOSOR110199-1GEA 992 CZK
APOSOR110199-1G APOSOR110199-5G
Ethyl 3,6-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-3,6-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Vzorec: C₁₀H₈Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02186226
CAS číslo: 478040-91-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR