Tisk…

3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile

Dodavatel: Apollo Scientific

Synonyms: 3-Bromo-6-cyanoimidazo[1,2-a]pyridine

OR110193-1G OR110193-5G
APOSOR110193-1GEA 3940 CZK
APOSOR110193-1G APOSOR110193-5G
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
Vzorec: C₈H₄BrN₃
Bod tání: 207…209 °C
Storage Temperature: Ambient
MDL Number: MFCD06659617
CAS číslo: 885950-21-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR