Tisk…

Ethyl-6-bromo-1-benzothiophene-2-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 6-bromo-1-benzothiophene-2-carboxylate

OR110187-1G OR110187-5G
APOSOR110187-1GEA 827 CZK
APOSOR110187-1G APOSOR110187-5G
Ethyl-6-bromo-1-benzothiophene-2-carboxylate
Ethyl-6-bromo-1-benzothiophene-2-carboxylate
Vzorec: C₁₁H₉BrO₂S
MW: 285,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100112
CAS číslo: 105191-64-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR