1-Methyl-6-nitro-1H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
OR110173-1G
OR110173-5G
OR110173-25G
APOSOR110173-1GEA
622
CZK
APOSOR110173-1G
APOSOR110173-5G
APOSOR110173-25G
1-Methyl-6-nitro-1H-indazole
1-Methyl-6-nitro-1H-indazole
Vzorec:
C₈H₇N₃O₂ MW: 177,160 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00456124 CAS číslo: 6850-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...