Tisk…

3-(2-Oxoimidazolidin-1-yl)benzoic acid

Dodavatel: Apollo Scientific
Varování

OR110169-1G OR110169-5G
APOSOR110169-1GEA 3310 CZK
APOSOR110169-1G APOSOR110169-5G
3-(2-Oxoimidazolidin-1-yl)benzoic acid
3-(2-Oxoimidazolidin-1-yl)benzoic acid
Vzorec: C₁₀H₁₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD05864567
CAS číslo: 884504-86-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR