8-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
8-Bromo-6-cyanoimidazo[1,2-a]pyridine
OR110151-1G
OR110151-250MG
OR110151-5G
APOSOR110151-1GEA
2780
CZK
APOSOR110151-1G
APOSOR110151-250MG
APOSOR110151-5G
8-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
8-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
Vzorec:
C₈H₄BrN₃ MW: 222,05 g/mol Bod tání: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD14584821 CAS číslo: 1221791-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...