Tisk…

4-Chloro-N-phenylbutyramide

Dodavatel: Apollo Scientific

Synonyms: 4-Chloro-N-phenylbutanamide

OR110143-10G OR110143-1G OR110143-5G
APOSOR110143-10GEA 0 CZK
APOSOR110143-10G APOSOR110143-1G APOSOR110143-5G
4-Chloro-N-phenylbutyramide
4-Chloro-N-phenylbutyramide
Vzorec: C₁₀H₁₂ClNO
MW: 197.662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01357318
CAS číslo: 7578-45-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR