5-tert-Butyl-2-iodo-1,3-dimethylbenzene
Dodavatel: Apollo Scientific
OR110132-10G
OR110132-1G
OR110132-5G
APOSOR110132-10GEA
13030
CZK
APOSOR110132-10G
APOSOR110132-1G
APOSOR110132-5G
5-tert-Butyl-2-iodo-1,3-dimethylbenzene
5-tert-Butyl-2-iodo-1,3-dimethylbenzene
Vzorec:
C₁₂H₁₇I MW: 288,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052173 CAS číslo: 5122-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...