Furan-3-boronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(3-Furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan
OR11010-25G
APOSOR11010-25GEA
6190
CZK
APOSOR11010-25G
Furan-3-boronic acid pinacol ester
Furan-3-boronic acid pinacol ester
Vzorec:
C₁₀H₁₅BO₃ MW: 194,035 g/mol Bod tání: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD03453669 CAS číslo: 248924-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...