Tisk…

4-Cyanophenylboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Cyanobenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyanophenylboronic acid, pinacol ester

OR11009-25G
APOSOR11009-25GEA 1990 CZK
APOSOR11009-25G
4-Cyanophenylboronic acid pinacol ester 98%
4-Cyanophenylboronic acid pinacol ester
Vzorec: C₁₃H₁₆BNO₂
MW: 229,09 g/mol
Bod tání: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD03093897
CAS číslo: 171364-82-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR