4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone
Dodavatel: Apollo Scientific
OR110076-1G
OR110076-5G
APOSOR110076-1GEA
7940
CZK
APOSOR110076-1G
APOSOR110076-5G
4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone
4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone
Vzorec:
C₈H₁₀Br₂N₂O MW: 310 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12079183 CAS číslo: 107784-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...