Tisk…

3-Aminophenylboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester

OR11005-25G
APOSOR11005-25GEA 2070 CZK
APOSOR11005-25G
3-Aminophenylboronic acid pinacol ester
3-Aminophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₈BNO₂
MW: 219,09 g/mol
Bod tání: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD03453668
CAS číslo: 210907-84-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR