5-Amino-3-(2-thienyl)-1H-pyrazole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Thien-2-yl-1H-pyrazol-3-amine
OR110021-1G
OR110021-5G
OR110021-10G
APOSOR110021-1GEA
910
CZK
APOSOR110021-1G
APOSOR110021-5G
APOSOR110021-10G
5-Amino-3-(2-thienyl)-1H-pyrazole
5-Amino-3-(2-thienyl)-1H-pyrazole
Vzorec:
C₇H₇N₃S MW: 165,21 g/mol Bod tání: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051815 CAS číslo: 96799-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...