Tisk…

Methyl-2-(p-tolyl)benzoate

Dodavatel: Apollo Scientific

Synonyms: Methyl 2-(4-Methylphenyl)benzoate, 4'-Methylbiphenyl-2-carboxylic Acid Methyl Ester, Methyl 4'-methyl-[1,1'-biphenyl]-2-carboxylate, Methyl 4'-Methylbiphenyl-2-carboxylate, 2-(p-Tolyl)benzoic Acid Methyl Ester

OR10997-25G
APOSOR10997-25GEA 4860 CZK
APOSOR10997-25G
Methyl-2-(p-tolyl)benzoate
Methyl-2-(p-tolyl)benzoate
Vzorec: C₁₅H₁₄O₂
MW: 226,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03453661
CAS číslo: 114772-34-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR