5-(tert-Butoxycarbonylamino)-1-pentanol 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-(5-Hydroxypentyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(5-Hydroxypentyl)carbamate, tert-Butyl (5-hydroxypentyl)carbamate, N-Boc-5-aminopentanol, 5-(Boc-amino)-1-pentanol
OR1099-1G
OR1099-25G
OR1099-5G
APOSOR1099-1GEA
1450
CZK
APOSOR1099-1G
APOSOR1099-25G
APOSOR1099-5G
5-(tert-Butoxycarbonylamino)-1-pentanol 95%
5-(tert-Butoxycarbonylamino)-1-pentanol
Vzorec:
C₁₀H₂₁NO₃ MW: 203,28 g/mol Hustota: 1 g/cm³ (25 °C) Bod vzplanutí: 109 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01862955 CAS číslo: 75178-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...