1,14-Tetradecanediol
Dodavatel: Apollo Scientific
Synonyms:
1,14-Dihydroxy tetradecane, Tetradecane-1,14-diol
OR10979-100G
APOSOR10979-100GEA
6750
CZK
APOSOR10979-100G
1,14-Tetradecanediol
1,14-Tetradecanediol
Vzorec:
C₁₄H₃₀O₂ MW: 230,39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004758 CAS číslo: 19812-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...