Tisk…

2-Nitrophenyl selenocyanate

Dodavatel: Apollo Scientific
Nebezpečí

OR10971-1G OR10971-5G OR10971-25G
APOSOR10971-1GEA 1580 CZK
APOSOR10971-1G APOSOR10971-5G APOSOR10971-25G
2-Nitrophenyl selenocyanate
2-Nitrophenyl selenocyanate
Vzorec: C₇H₄N₂O₂Se
MW: 227.092 g/mol
Bod tání: 134…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00043146
CAS číslo: 51694-22-5
EINECS: 257-350-4
UN: 3283
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR