Tisk…

S-Methylisothiosemicarbazide hydroiodide

Dodavatel: Apollo Scientific
Varování

OR10968-1G OR10968-5G
APOSOR10968-1GEA 622 CZK
APOSOR10968-1G APOSOR10968-5G
S-Methylisothiosemicarbazide hydroiodide
S-Methylisothiosemicarbazide hydroiodide
Vzorec: C₂H₈IN₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00035462
CAS číslo: 35600-34-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR