Tisk…

2-Aminobenzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Aminophenyl)methanol, 2-Hydroxymethylaniline, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl Alcohol

OR10943-250G OR10943-50G
APOSOR10943-250GEA 3310 CZK
APOSOR10943-250G APOSOR10943-50G
2-Aminobenzyl alcohol
2-Aminobenzyl alcohol
Vzorec: C₇H₉NO
MW: 123,15 g/mol
Bod varu: 160 °C/10mmHg
Bod tání: 81…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS číslo: 5344-90-1
EINECS: 226-293-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR