3-Ethoxyphenylboronic acid 98%
Dodavatel: Apollo Scientific
OR10932-100G
OR10932-25G
APOSOR10932-100GEA
15440
CZK
APOSOR10932-100G
APOSOR10932-25G
3-Ethoxyphenylboronic acid 98%
3-Ethoxyphenylboronic acid
Vzorec:
C₈H₁₁BO₃ MW: 165,98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00274219 CAS číslo: 90555-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...