Tisk…

Hydrochinonmonomethylether 98+%

Dodavatel: Apollo Scientific
Varování

Synonyms: p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole

OR10915-1KG OR10915-250G
APOSOR10915-1KGEA 2240 CZK
APOSOR10915-1KG APOSOR10915-250G
Hydrochinonmonomethylether 98+%
Hydrochinonmonomethylether
Vzorec: C₇H₈O₂
MW: 124.14
Bod varu: 243 °C
Bod tání: 55…57 °C
Hustota: 1.55
Bod vzplanutí: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS číslo: 150-76-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR