Tisk…

4-Ethoxychrysoidine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(4-Ethoxyphenylazo)-1,3-phenylenediamine Monohydrochloride, Ethoxy Red, 4-(4-Ethoxyphenylazo)-m-phenylenediamine

OR10870-10G OR10870-1G OR10870-5G
APOSOR10870-10GEA 6420 CZK
APOSOR10870-10G APOSOR10870-1G APOSOR10870-5G
4-Ethoxychrysoidine hydrochloride
4-Ethoxychrysoidine hydrochloride
Vzorec: C₁₄H₁₆N₄O
MW: 292,77 g/mol
Bod tání: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00042023
CAS číslo: 2313-87-3
EINECS: 219-010-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR