2-(Ethanesulphonylamino)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR10869-1G
OR10869-500MG
APOSOR10869-1GEA
7670
CZK
APOSOR10869-1G
APOSOR10869-500MG
2-(Ethanesulphonylamino)benzoic acid
2-(Ethanesulphonylamino)benzoic acid
Vzorec:
C₉H₁₁NO₄S MW: 229.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03411193 CAS číslo: 923248-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...