3-Acetyl-1-(phenylsulphonyl)pyrrole
Dodavatel: Apollo Scientific
OR10821-1G
OR10821-5G
APOSOR10821-1GEA
0
CZK
APOSOR10821-1G
APOSOR10821-5G
3-Acetyl-1-(phenylsulphonyl)pyrrole
3-Acetyl-1-(phenylsulphonyl)pyrrole
Vzorec:
C₁₂H₁₁NO₃S Storage Temperature: Ambient |
MDL Number:
MFCD00067752 CAS číslo: 81453-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...