Tisk…

(3-Methylbenzyl)phosphonic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR10786-1G OR10786-5G
APOSOR10786-1GEA 3310 CZK
APOSOR10786-1G APOSOR10786-5G
(3-Methylbenzyl)phosphonic acid
(3-Methylbenzyl)phosphonic acid
Vzorec: C₈H₁₁O₃P
Bod tání: 194…195 °C
Storage Temperature: Ambient
CAS číslo: 18945-65-8
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR