Tisk…

Diethyl (hydroxymethyl)phosphonate 97%

Dodavatel: Apollo Scientific
Varování

OR10715-100G OR10715-25G
APOSOR10715-100GEA 1580 CZK
APOSOR10715-100G APOSOR10715-25G
Diethyl (hydroxymethyl)phosphonate 97%
Diethyl (hydroxymethyl)phosphonate
Vzorec: C₅H₁₃O₄P
Bod varu: 124…126 °C/3mmHg
Hustota: 1,14 g/cm³ (25 °C)
Bod vzplanutí: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00014418
CAS číslo: 3084-40-0
EINECS: 221-391-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR