Tisk…

3-(Bromoacetyl)pyridine ydrobromide

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(2-Bromoacetyl)pyridine hydrobromide

OR10712-1G OR10712-25G OR10712-5G
APOSOR10712-1GEA 622 CZK
APOSOR10712-1G APOSOR10712-25G APOSOR10712-5G
3-(Bromoacetyl)pyridine ydrobromide
3-(Bromoacetyl)pyridine ydrobromide
Vzorec: C₇H₆BrNO
MW: 280,95 g/mol
Bod tání: 192…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00052182
CAS číslo: 17694-68-7
EINECS: 241-692-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR