Triethyl phosphonoacetate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate
OR10710-100G
OR10710-1KG
OR10710-500G
APOSOR10710-100GEA
827
CZK
APOSOR10710-100G
APOSOR10710-1KG
APOSOR10710-500G
Triethyl phosphonoacetate
Triethyl phosphonoacetate
Vzorec:
C₈H₁₇O₅P MW: 224,19 g/mol Bod varu: 260…262 °C Bod tání: –24 °C Hustota: 1,13 g/cm³ (25 °C) Bod vzplanutí: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009177 CAS číslo: 867-13-0 EINECS: 212-757-6 UN: 3082 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...