Tisk…

tert-Butyl (diethylphosphono)acetate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 2-(diethoxyphosphoryl)acetate, Diethyl(2-tert-butoxycarbonylmethyl)phosphonate

OR10694-100G OR10694-1KG OR10694-500G
APOSOR10694-100GEA 3110 CZK
APOSOR10694-100G APOSOR10694-1KG APOSOR10694-500G
tert-Butyl (diethylphosphono)acetate 98%
tert-Butyl (diethylphosphono)acetate
Vzorec: C₁₀H₂₁O₅P
Bod varu: 100…103 °C/1,5mmHg
Hustota: 1,074 g/cm³ (25 °C)
Bod vzplanutí: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075414
CAS číslo: 27784-76-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR