Tisk…

1,4-Butylenediphosphonic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1,4-Butanediphosphonic acid, 1,4-Butanediphosphonicacid

OR10676-1G OR10676-250MG
APOSOR10676-1GEA 1660 CZK
APOSOR10676-1G APOSOR10676-250MG
1,4-Butylenediphosphonic acid
1,4-Butylenediphosphonic acid
Vzorec: C₄H₁₂O₆P₂
MW: 218,0820 g/mol
Bod tání: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD01631212
CAS číslo: 4671-77-6
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR