Tisk…

1,4-Phenylenediphosphonic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1,4-Benzenediphosphonic acid

OR10672-1G OR10672-250MG
APOSOR10672-1GEA 3310 CZK
APOSOR10672-1G APOSOR10672-250MG
1,4-Phenylenediphosphonic acid
1,4-Phenylenediphosphonic acid
Vzorec: C₆H₈O₆P₂
MW: 238,07 g/mol
Bod tání: >260 °C
Storage Temperature: Ambient
MDL Number: MFCD00475582
CAS číslo: 880-68-2
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR