Tisk…

N-Boc-4-bromoaniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromoaniline, N-BOC protected, tert-Butyl (4-bromophenyl)carbamate

OR10620-25G OR10620-5G OR10620-1G
APOSOR10620-25GEA 2480 CZK
APOSOR10620-25G APOSOR10620-5G APOSOR10620-1G
N-Boc-4-bromoaniline
N-Boc-4-bromoaniline
Vzorec: C₁₁H₁₄BrNO₂
Bod tání: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD01006612
CAS číslo: 131818-17-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR