Tisk…

Bis(hexylene glycolato)diboron

Dodavatel: Apollo Scientific
Varování

Synonyms: 4,4,4',4',6,6'-Hexamethyl-2,2'-bi-1,3,2-dioxaborinane, 4,4,4',4',6,6'-Hexamethyl-2,2'-bi(1,3,2-dioxaborinane)

OR10606-25G OR10606-100G
APOSOR10606-25GEA 1700 CZK
APOSOR10606-25G APOSOR10606-100G
Bis(hexylene glycolato)diboron
Bis(hexylene glycolato)diboron
Vzorec: C₁₂H₂₄B₂O₄
MW: 253,94 g/mol
Bod tání: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD06246008
CAS číslo: 230299-21-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR