Tisk…

4-Bromo-2-chlorobenzoic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR1060-25G OR1060-100G OR1060-5G
APOSOR1060-25GEA 1290 CZK
APOSOR1060-25G APOSOR1060-100G APOSOR1060-5G
4-Bromo-2-chlorobenzoic acid
4-Bromo-2-chlorobenzoic acid
Vzorec: C₇H₄BrClO₂
MW: 235,46 g/mol
Bod tání: 171…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040903
CAS číslo: 59748-90-2
EINECS: 261-911-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR