Tisk…

4-Aminonicotinic acid 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Amino-3-pyridinecarboxylic acid, 4-Amino-nicotinic acid, 4-Amino-3-pyridinecarboxylic acid

OR1058-1G OR1058-25G OR1058-5G
APOSOR1058-1GEA 622 CZK
APOSOR1058-1G APOSOR1058-25G APOSOR1058-5G
4-Aminonicotinic acid 99%
4-Aminonicotinic acid
Vzorec: C₆H₆N₂O₂
Bod tání: 307…312 °C
Storage Temperature: Ambient
MDL Number: MFCD00234183
CAS číslo: 7418-65-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR