2-(Methylthio)phenylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-(Methylthio)phenyl)boronic acid, 2-(Methylthio)benzeneboronic acid
OR10566-100G
OR10566-25G
APOSOR10566-100GEA
8570
CZK
APOSOR10566-100G
APOSOR10566-25G
2-(Methylthio)phenylboronic acid 97%
2-(Methylthio)phenylboronic acid
Vzorec:
C₇H₉BO₂S Bod tání: 77…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318165 CAS číslo: 168618-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...