3-(Methylcarbamoyl)phenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(N-Methylaminocarbonyl)phenylboronic acid, 3-(Methylcarbamoyl)benzeneboronic acid, (3-(Methylcarbamoyl)phenyl)boronic acid
OR10554-1G
OR10554-250MG
APOSOR10554-1GEA
910
CZK
APOSOR10554-1G
APOSOR10554-250MG
3-(Methylcarbamoyl)phenylboronic acid
3-(Methylcarbamoyl)phenylboronic acid
Vzorec:
C₈H₁₀BNO₃ MW: 178,98 g/mol Bod tání: 114…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038918 CAS číslo: 832695-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...