4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
OR10537-100G
APOSOR10537-100GEA
5230
CZK
APOSOR10537-100G
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Vzorec:
C₁₄H₁₉BO₄ MW: 262,11 g/mol Bod tání: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179438 CAS číslo: 171364-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...