3-Methoxycarbonyl-5-nitrobenzeneboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, 3-Methoxycarbonyl-5-nitrophenylboronic acid, 3-(Methoxycarbonyl)-5-nitrophenylboronic acid, (3-(Methoxycarbonyl)-5-nitrophenyl)boronic acid
OR10532-25G
APOSOR10532-25GEA
4440
CZK
APOSOR10532-25G
3-Methoxycarbonyl-5-nitrobenzeneboronic acid
3-Methoxycarbonyl-5-nitrobenzeneboronic acid
Vzorec:
C₈H₈BNO₆ MW: 224,96 g/mol Bod tání: 250…255 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179469 CAS číslo: 117342-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...