4-Methoxy-3-methylphenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Methoxy-3-methylbenzeneboronic acid
OR10528-100G
OR10528-25G
OR10528-5G
APOSOR10528-100GEA
16960
CZK
APOSOR10528-100G
APOSOR10528-25G
APOSOR10528-5G
4-Methoxy-3-methylphenylboronic acid 98%
4-Methoxy-3-methylphenylboronic acid
Vzorec:
C₈H₁₁BO₃ MW: 165,99 g/mol Bod tání: 194…200 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179464 CAS číslo: 175883-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...