Tisk…

4-(Isopropylcarbamoyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(N-Isopropylaminocarbonyl)phenylboronic acid, (4-(Isopropylcarbamoyl)phenyl)boronic acid, 4-(Isopropylcarbamoyl)benzeneboronic acid

OR10521-1G OR10521-5G
APOSOR10521-1GEA 2150 CZK
APOSOR10521-1G APOSOR10521-5G
4-(Isopropylcarbamoyl)phenylboronic acid
4-(Isopropylcarbamoyl)phenylboronic acid
Vzorec: C₁₀H₁₄BNO₃
MW: 207,04 g/mol
Bod tání: 162…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039353
CAS číslo: 397843-67-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR