4-Hydroxyphenylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
OR10505-25G
OR10505-5G
OR10505-100G
APOSOR10505-25GEA
992
CZK
APOSOR10505-25G
APOSOR10505-5G
APOSOR10505-100G
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid
Vzorec:
C₆H₇BO₃ MW: 137,93 g/mol Bod tání: 285 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD01074628 CAS číslo: 71597-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...