Tisk…

2-Hydroxyphenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydroxybenzeneboronic acid, Dihydroxy(2-hydroxyphenyl)borane, (2-Hydroxyphenyl)metaboric acid

OR10503-25G OR10503-5G OR10503-100G
APOSOR10503-25GEA 1700 CZK
APOSOR10503-25G APOSOR10503-5G APOSOR10503-100G
2-Hydroxyphenylboronic acid
2-Hydroxyphenylboronic acid
Vzorec: C₆H₇BO₃
MW: 137,93 g/mol
Bod tání: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074581
CAS číslo: 89466-08-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR