Tisk…

4-(Ethylsulphonyl)phenylboronic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-(Ethylsulphonyl)phenyl)boronic acid, 4-(Ethylsulphonyl)benzeneboronic acid

OR10479-1G OR10479-500MG
APOSOR10479-1GEA 3740 CZK
APOSOR10479-1G APOSOR10479-500MG
4-(Ethylsulphonyl)phenylboronic acid 97%
4-(Ethylsulphonyl)phenylboronic acid
Vzorec: C₈H₁₁BO₄S
MW: 214,05 g/mol
Bod tání: 135…139 °C
Storage Temperature: Ambient
MDL Number: MFCD02093076
CAS číslo: 352530-24-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR