Ethyl-2-aminothiazole-4-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 2-aminothiazole-4-carboxylate
OR10473-25G
OR10473-5G
APOSOR10473-25GEA
705
CZK
APOSOR10473-25G
APOSOR10473-5G
Ethyl-2-aminothiazole-4-carboxylate
Ethyl-2-aminothiazole-4-carboxylate
Vzorec:
C₆H₈N₂O₂S MW: 172,21 g/mol Bod tání: 175…181 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00619079 CAS číslo: 5398-36-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...