Tisk…

4-Ethoxyphenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Ethoxybenzeneboronic acid, (4-Ethoxyphenyl)metaboric acid, Dihydroxy(4-ethoxyphenyl)borane

OR10472-100G
APOSOR10472-100GEA 3640 CZK
APOSOR10472-100G
4-Ethoxyphenylboronic acid
4-Ethoxyphenylboronic acid
Vzorec: C₈H₁₁BO₃
MW: 165,98 g/mol
Bod tání: 140…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00674028
CAS číslo: 22237-13-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR