2-Ethoxyphenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR10471-100G
OR10471-25G
APOSOR10471-100GEA
7380
CZK
APOSOR10471-100G
APOSOR10471-25G
2-Ethoxyphenylboronic acid
2-Ethoxyphenylboronic acid
Vzorec:
C₈H₁₁BO₃ Bod tání: 97…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674027 CAS číslo: 213211-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...