Tisk…

4-(Ethoxycarbonyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

OR10470-100G OR10470-25G OR10470-5G
APOSOR10470-100GEA 4200 CZK
APOSOR10470-100G APOSOR10470-25G APOSOR10470-5G
4-(Ethoxycarbonyl)phenylboronic acid
4-(Ethoxycarbonyl)phenylboronic acid
Vzorec: C₉H₁₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD02179441
CAS číslo: 4334-88-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR