Tisk…

3-(Ethoxycarbonyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

OR10469-25G OR10469-5G OR10469-100G
APOSOR10469-25GEA 992 CZK
APOSOR10469-25G APOSOR10469-5G APOSOR10469-100G
3-(Ethoxycarbonyl)phenylboronic acid
3-(Ethoxycarbonyl)phenylboronic acid
Vzorec: C₉H₁₁BO₄
MW: 193,99 g/mol
Bod tání: 131…139 °C
Storage Temperature: Ambient
MDL Number: MFCD02179444
CAS číslo: 4334-87-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR