4-tert-Butylphenylboronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-tert-Butylbenzeneboronic acid, (4-(tert-Butyl)phenyl)boronic acid
OR10399-100G
OR10399-25G
APOSOR10399-100GEA
1740
CZK
APOSOR10399-100G
APOSOR10399-25G
4-tert-Butylphenylboronic acid 98%
4-tert-Butylphenylboronic acid
Vzorec:
C₁₀H₁₅BO₂ MW: 178,04 g/mol Bod tání: 191…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD01009697 CAS číslo: 123324-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...